Assay Details
Gene Symbol: HS1BP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710053.3 | XP_002710099.1 | 4 - 5 | 710 | 67 | |
XM_017340907.1 | XP_017196396.1 | 4 - 5 | 710 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
