Assay Details
| Gene Symbol: TSHZ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008261472.2 | XP_008259694.1 | 1 - 2 | 298 | 76 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008261472.2 transcripts.
Set Membership: |
|

TSHZ1
Genomic Map