Assay Details
| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347500.1 | XP_017202989.1 | 3 | 1372 | 120 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

NPFFR2| Gene Symbol: NPFFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017347500.1 | XP_017202989.1 | 3 | 1372 | 120 | |
Set Membership: |
|