Assay Details
Gene Symbol: SAPCD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714311.3 | XP_002714357.1 | 1 - 2 | 340 | 100 | |
XM_008262592.2 | XP_008260814.1 | 1 - 2 | 340 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
