Assay Details
Gene Symbol: CRPPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720852.3 | XP_002720898.1 | 932 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
