Assay Details
| Gene Symbol: CCL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249145.2 | XP_008247367.1 | 305 | 66 | ||
| XM_017337982.1 | XP_017193471.1 | 305 | 66 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCL25| Gene Symbol: CCL25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008249145.2 | XP_008247367.1 | 305 | 66 | ||
| XM_017337982.1 | XP_017193471.1 | 305 | 66 | |||
Set Membership: |
|