Assay Details
| Gene Symbol: PPARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723354.3 | XP_002723400.1 | 1000 | 72 | ||
| GenBank mRNA | AF013264.1 | - | 64 | 72 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPARA| Gene Symbol: PPARA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723354.3 | XP_002723400.1 | 1000 | 72 | ||
| GenBank mRNA | AF013264.1 | - | 64 | 72 | ||
Set Membership: |
|