Assay Details
| Gene Symbol: RLA-DRB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714606.3 | XP_002714652.2 | 2 | 301 | 60 | |
| GenBank mRNA | EU427425.1 | - | 1 | 118 | 60 | |
More Information
Set Membership: |
|

RLA-DRB1
Genomic Map