Assay Details
| Gene Symbol: NQO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720966.3 | XP_002721012.1 | 621 | 61 | ||
| XM_008274279.2 | XP_008272501.1 | 500 | 61 | |||
| GenBank mRNA | FJ474089.1 | - | 387 | 61 | ||
More Information
Set Membership: |
|

NQO2
Genomic Map