Assay Details
| Gene Symbol: MXRA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017338916.1 | XP_017194405.1 | 464 | 95 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MXRA7| Gene Symbol: MXRA7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017338916.1 | XP_017194405.1 | 464 | 95 | ||
Set Membership: |
|