Assay Details
Gene Symbol: POPDC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002714844.3 | XP_002714890.1 | 2 - 3 | 1067 | 90 | |
XM_008263328.2 | XP_008261550.1 | 3 - 4 | 1168 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
