Assay Details
| Gene Symbol: UPRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721822.3 | XP_002721868.1 | 918 | 84 | ||
| XM_008248849.2 | XP_008247071.1 | 921 | 84 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

UPRT| Gene Symbol: UPRT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721822.3 | XP_002721868.1 | 918 | 84 | ||
| XM_008248849.2 | XP_008247071.1 | 921 | 84 | |||
Set Membership: |
|