Assay Details
| Gene Symbol: MAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263854.2 | XP_008262076.1 | 1 - 2 | 2240 | 74 | |
| XM_008263864.2 | XP_008262086.1 | 1 - 2 | 2240 | 74 | ||
More Information
Set Membership: |
|

MAP6
Genomic Map