Assay Details
Gene Symbol: HJV | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002715642.3 | XP_002715688.1 | 3 - 4 | 955 | 78 | |
XM_008264524.2 | XP_008262746.1 | 3 - 4 | 976 | 78 | ||
XM_008264525.2 | XP_008262747.1 | 2 - 3 | 767 | 78 | ||
XM_017345926.1 | XP_017201415.1 | 2 - 3 | 1989 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
