Assay Details
Gene Symbol: LOC100345657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008249573.2 | XP_008247795.1 | 21 - 22 | 3168 | 120 | |
XM_008249579.2 | XP_008247801.1 | 21 - 22 | 3099 | 120 | ||
XM_008249585.2 | XP_008247807.1 | 21 - 22 | 3162 | 120 | ||
XM_008249590.2 | XP_008247812.1 | 21 - 22 | 3162 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
