Assay Details
| Gene Symbol: PPP1R3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274068.1 | XP_008272290.1 | 219 | 79 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008274068.1 transcripts.
Set Membership: |
|

PPP1R3B
Genomic Map