Assay Details
| Gene Symbol: LOC100339265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718760.3 | XP_002718806.1 | 11 - 12 | 1343 | 112 | |
| XM_008270572.2 | XP_008268794.1 | 11 - 12 | 1247 | 112 | ||
More Information
Set Membership: |
|

LOC100339265
Genomic Map