Assay Details
| Gene Symbol: LOC100339265 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718760.3 | XP_002718806.1 | 4 - 5 | 493 | 66 | |
| XM_008270572.2 | XP_008268794.1 | 4 - 5 | 397 | 66 | ||
More Information
Set Membership: |
|

LOC100339265
Genomic Map