Assay Details
Gene Symbol: PPM1K | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717007.3 | XP_002717053.1 | 5 - 6 | 953 | 62 | |
XM_008267615.2 | XP_008265837.1 | 5 - 6 | 1127 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
