Assay Details
| Gene Symbol: ASB14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260830.2 | XP_008259052.2 | 8 - 9 | 2391 | 74 | |
| XM_017343894.1 | XP_017199383.1 | 7 - 8 | 2308 | 74 | ||
| XM_017343895.1 | XP_017199384.1 | 8 - 9 | 2361 | 74 | ||
| XM_017343896.1 | XP_017199385.1 | 7 - 8 | 2334 | 74 | ||
| XM_017343897.1 | XP_017199386.1 | 6 - 7 | 1305 | 74 | ||
More Information
Set Membership: |
|

ASB14
Genomic Map