Assay Details
| Gene Symbol: KEAP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002723276.3 | XP_002723322.1 | 2070 | 68 | ||
| XM_008251548.2 | XP_008249770.1 | 1938 | 68 | |||
| XM_008251549.2 | XP_008249771.1 | 1991 | 68 | |||
| XM_008251550.2 | XP_008249772.1 | 1961 | 68 | |||
More Information
Set Membership: |
|

KEAP1
Genomic Map