Assay Details
| Gene Symbol: MRPS22 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716552.3 | XP_002716598.1 | 2 - 3 | 379 | 62 | |
| XM_008266238.2 | XP_008264460.1 | 2 - 3 | 379 | 62 | ||
| XM_008266239.2 | XP_008264461.1 | 2 - 3 | 284 | 62 | ||
More Information
Set Membership: |
|

MRPS22
Genomic Map