Assay Details
| Gene Symbol: TM2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720774.3 | XP_002720820.1 | 416 | 67 | ||
| XM_008273958.2 | XP_008272180.1 | 547 | 67 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TM2D2| Gene Symbol: TM2D2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720774.3 | XP_002720820.1 | 416 | 67 | ||
| XM_008273958.2 | XP_008272180.1 | 547 | 67 | |||
Set Membership: |
|