Assay Details
Gene Symbol: TMED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711656.3 | XP_002711702.1 | 3 - 4 | 718 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
