Assay Details
| Gene Symbol: PIGU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710794.3 | XP_002710840.1 | 10 - 11 | 1098 | 63 | |
| XM_008256084.2 | XP_008254306.1 | 9 - 10 | 1091 | 63 | ||
More Information
Set Membership: |
|

PIGU
Genomic Map