Assay Details
| Gene Symbol: LOC100352550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008248917.2 | XP_008247139.1 | 87 | 60 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008248917.2 transcripts.
Set Membership: |
|

LOC100352550
Genomic Map