Assay Details
Gene Symbol: LOC100352550 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248916.2 | XP_008247138.1 | 866 | 93 | ||
XM_008248917.2 | XP_008247139.1 | 714 | 93 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
