Assay Details
| Gene Symbol: GRAMD2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710167.3 | XP_002710213.1 | 2 - 3 | 336 | 62 | |
| XM_017341016.1 | XP_017196505.1 | 2 - 3 | 339 | 62 | ||
| XM_017341017.1 | XP_017196506.1 | 2 - 3 | 329 | 62 | ||
More Information
Set Membership: |
|

GRAMD2B
Genomic Map