Assay Details
| Gene Symbol: GLRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008268724.2 | XP_008266946.1 | 2 - 3 | 239 | 59 | |
| XM_008268725.2 | XP_008266947.1 | 2 - 3 | 94 | 59 | ||
| XM_008268728.2 | XP_008266950.1 | 3 - 4 | 142 | 59 | ||
More Information
Set Membership: |
|

GLRX2
Genomic Map