Assay Details
Gene Symbol: TPI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712911.2 | XP_002712957.1 | 6 - 7 | 762 | 61 | |
XM_017343417.1 | XP_017198906.1 | 5 - 6 | 515 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
