Assay Details
| Gene Symbol: TUBA4A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712488.3 | XP_002712534.1 | 1 - 2 | 136 | 70 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002712488.3 transcripts.
Set Membership: |
|

TUBA4A
Genomic Map