Assay Details
Gene Symbol: NFE2L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008271359.2 | XP_008269581.1 | 5 - 6 | 1571 | 78 | |
XM_008271360.2 | XP_008269582.1 | 5 - 6 | 1559 | 78 | ||
XM_008271361.2 | XP_008269583.1 | 5 - 6 | 1624 | 78 | ||
XM_008271362.2 | XP_008269584.1 | 5 - 6 | 1585 | 78 | ||
XM_008271364.2 | XP_008269586.1 | 4 - 5 | 534 | 78 | ||
XM_017349171.1 | XP_017204660.1 | 4 - 5 | 1469 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
