Assay Details
Gene Symbol: LOC100341681 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008257045.2 | XP_008255267.1 | 3 - 4 | 219 | 70 | |
XM_017342096.1 | XP_017197585.1 | 3 - 4 | 219 | 70 | ||
XM_017342097.1 | XP_017197586.1 | 3 - 4 | 219 | 70 | ||
XM_017342098.1 | XP_017197587.1 | 3 - 4 | 219 | 70 | ||
XM_017342099.1 | XP_017197588.1 | 3 - 4 | 219 | 70 | ||
XM_017342100.1 | XP_017197589.1 | 3 - 4 | 219 | 70 | ||
XM_017342101.1 | XP_017197590.1 | 3 - 4 | 219 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
