Assay Details
Gene Symbol: MAP2K4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718920.3 | XP_002718966.1 | 7 - 8 | 883 | 56 | |
XM_017348962.1 | XP_017204451.1 | 8 - 9 | 918 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
