Assay Details
| Gene Symbol: ZXDC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713143.3 | XP_002713189.1 | 1 - 2 | 1118 | 55 | |
| XM_008260309.2 | XP_008258531.1 | 1 - 2 | 1120 | 55 | ||
More Information
Set Membership: |
|

ZXDC
Genomic Map