Assay Details
Gene Symbol: POU6F1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008256468.2 | XP_008254690.1 | 11 - 12 | 2138 | 54 | |
XM_017341860.1 | XP_017197349.1 | 11 - 12 | 2260 | 54 | ||
XM_017341861.1 | XP_017197350.1 | 12 - 13 | 2326 | 54 | ||
XM_017341862.1 | XP_017197351.1 | 11 - 12 | 2283 | 54 | ||
XM_017341863.1 | XP_017197352.1 | 9 - 10 | 1834 | 54 | ||
XM_017341864.1 | XP_017197353.1 | 10 - 11 | 2064 | 54 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
