Assay Details
| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724146.2 | XP_002724192.1 | 357 | 65 | ||
| XM_008253294.2 | XP_008251516.1 | 393 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMEM205| Gene Symbol: TMEM205 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724146.2 | XP_002724192.1 | 357 | 65 | ||
| XM_008253294.2 | XP_008251516.1 | 393 | 65 | |||
Set Membership: |
|