Assay Details
| Gene Symbol: GLOD4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017348914.1 | XP_017204403.1 | 5 - 6 | 535 | 56 | |
| XM_017348915.1 | XP_017204404.1 | 4 - 5 | 498 | 56 | ||
More Information
Set Membership: |
|

GLOD4
Genomic Map