Assay Details
| Gene Symbol: CCNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724206.3 | XP_002724252.3 | 625 | 61 | ||
| XM_008253463.2 | XP_008251685.2 | 625 | 61 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCNL2| Gene Symbol: CCNL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002724206.3 | XP_002724252.3 | 625 | 61 | ||
| XM_008253463.2 | XP_008251685.2 | 625 | 61 | |||
Set Membership: |
|