Assay Details
Gene Symbol: LGALS3BP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002724338.3 | XP_002724384.1 | 370 | 67 | ||
XM_017340531.1 | XP_017196020.1 | 359 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
