Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008254501.2 | XP_008252723.1 | 1 - 2 | 199 | 83 | |
XM_008254505.2 | XP_008252727.1 | 1 - 2 | 197 | 83 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008254501.2, XM_008254505.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
