Assay Details
Gene Symbol: GEMIN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002709909.3 | XP_002709955.1 | 2 - 3 | 281 | 102 | |
XM_008254505.2 | XP_008252727.1 | 2 - 3 | 338 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
