Assay Details
| Gene Symbol: LCLAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709935.3 | XP_002709981.1 | 6 | 4415 | 67 | |
| XM_008254571.2 | XP_008252793.1 | 7 | 4578 | 67 | ||
| XM_008254572.2 | XP_008252794.1 | 7 | 4656 | 67 | ||
| XM_008254574.2 | XP_008252796.1 | 6 | 4378 | 67 | ||
| XM_017340837.1 | XP_017196326.1 | 7 | 4611 | 67 | ||
| XM_017340838.1 | XP_017196327.1 | 7 | 4443 | 67 | ||
| XM_017340839.1 | XP_017196328.1 | 6 | 4358 | 67 | ||
| XM_017340840.1 | XP_017196329.1 | 6 | 4274 | 67 | ||
More Information
Set Membership: |
|

LCLAT1
Genomic Map