Assay Details
| Gene Symbol: TMEM170A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008257626.2 | XP_008255848.1 | 2 - 3 | 432 | 63 | |
| XM_017342367.1 | XP_017197856.1 | 2 - 3 | 488 | 63 | ||
| XM_017342369.1 | XP_017197858.1 | 2 - 3 | 282 | 63 | ||
More Information
Set Membership: |
|

TMEM170A
Genomic Map