Assay Details
Gene Symbol: LOC100347443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008275338.2 | XP_008273560.1 | 187 | 120 | ||
XM_017337551.1 | XP_017193040.1 | 187 | 120 |

More Information
Set Membership: |
![]() ![]() |
