Assay Details
Gene Symbol: LOC100347443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008275337.2 | XP_008273559.2 | 339 | 96 | ||
XM_008275338.2 | XP_008273560.1 | 394 | 96 | |||
XM_017337551.1 | XP_017193040.1 | 487 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008275337.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
