Assay Details
Gene Symbol: LOC100347443 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008275337.2 | XP_008273559.2 | 712 | 109 | ||
XM_008275338.2 | XP_008273560.1 | 767 | 109 | |||
XM_017337551.1 | XP_017193040.1 | 860 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() |
