Assay Details
Gene Symbol: LOC100343730 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017345680.1 | XP_017201169.1 | 9 - 10 | 973 | 108 | |
XM_017345681.1 | XP_017201170.1 | 9 - 10 | 973 | 108 | ||
XM_017345682.1 | XP_017201171.1 | 9 - 10 | 961 | 108 | ||
XM_017345683.1 | XP_017201172.1 | 8 - 9 | 952 | 108 | ||
XM_017345684.1 | XP_017201173.1 | 8 - 9 | 940 | 108 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
