Assay Details
Gene Symbol: NIF3L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002712443.3 | XP_002712489.1 | 2 - 3 | 562 | 70 | |
XM_017343014.1 | XP_017198503.1 | 3 - 4 | 639 | 70 | ||
XM_017343015.1 | XP_017198504.1 | 3 - 4 | 607 | 70 | ||
XM_017343016.1 | XP_017198505.1 | 3 - 4 | 639 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
