Assay Details
Gene Symbol: PAK4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002711469.3 | XP_002711515.1 | 4 - 5 | 1306 | 57 | |
XM_008257149.2 | XP_008255371.1 | 5 - 6 | 1380 | 57 | ||
XM_008257150.2 | XP_008255372.1 | 6 - 7 | 1491 | 57 | ||
XM_008257151.2 | XP_008255373.1 | 5 - 6 | 1405 | 57 | ||
XM_008257152.2 | XP_008255374.1 | 5 - 6 | 1330 | 57 | ||
XM_008257153.2 | XP_008255375.1 | 3 - 4 | 1407 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
